Learn More
Description
Specifications
Specifications
| Preparation Method | H2O (1mg/mL) or Aqueous buffers (1mg/mL) |
| Quantity | 5 kU |
| Source | Rhizopus sp. |
| Immunogen | Glucoamylase |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C. Avoid freeze/thaw cycles of solutions. |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.