Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glucose-6-phosphate Dehydrogenase, Leuconostoc mesenteroides, Protein

Catalog No. 346774500U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-677-4500U 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-677-4500U Supplier MilliporeSigma™ Supplier No. 346774500U
Add to Cart
Add to Cart
Native glucose-6-phosphate dehydrogenase from Leuconostoc mesenteroides. Catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconate. The enzyme can use either NAD+ or NADP+ as an electron acceptor. Reaction velocity with NAD+ is ∽1.8 times greater than that with NADP+. Note: 10,000 NADP units are equivalent to ∽18,000 NAD units.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Reconstitute to 1mg/mL in 5mM glycine buffer, pH 8.0. Agitation of solution may cause precipitation.
Quantity 500 U
Source Leuconostoc mesenteroides
Immunogen Glucose-6-phosphate Dehydrogenase
Storage Requirements Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.
Regulatory Status RUO
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.