Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ β-Glucuronidase, Helix pomatia, Protein

Catalog No. 3474201MU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-742-01MU 1 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-742-01MU Supplier MilliporeSigma™ Supplier No. 3474201MU
Add to Cart
Add to Cart
Native β-glucuronidase from Helix pomatia. Useful for hydrolyzing urinary steroid conjugates prior to steriod assays. Note: 1MU = 1,000,000 units.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 65% saturated ammonium sulfate suspension.
Preparation Method aqueous buffers
Quantity 1 mU
Source Helix pomatia
Immunogen β-Glucuronidase
Storage Requirements Following reconstitution, store in refrigerator (4°C). Stock solutions are stable for up to 3 months at 4°C.
Regulatory Status RUO
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.