Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Granulocyte Macrophage Colony-Stimulating Factor, Mouse, Recombinant Protein, E. coli

Catalog No. 23437410UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-437-410UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-437-410UG Supplier MilliporeSigma™ Supplier No. 23437410UG
Add to Cart
Add to Cart
Recombinant, mouse granulocyte macrophage colony-stimulating factor expressed in E. coli. Pleiotropic cytokine that can stimulate the proliferation, maturation, and function of hematopoietic cells. Reported to improve the response rate to antibiotic therapy in cancer subjects. Activates JAK2 in a variety of cells.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered PBS containing 50μg BSA/μg GM-CSF.
Preparation Method Reconstitute to a concentration of ≥10μg/mL with sterile PBS containing ≥0.1% HSA or BSA.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen Granulocyte Macrophage Colony-Stimulating Factor
Storage Requirements Following reconstitution, aliquot and freeze (−70°C) for long term storage or refrigerate (4°C) for short term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for 3 months at 4°C.
Regulatory Status RUO
Species Mouse
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.