Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Growth Hormone, Human Pituitary, Iodination Grade, Protein

Catalog No. 86900850UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-900-850UG 50 μg
86-900-8100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-900-850UG Supplier MilliporeSigma™ Supplier No. 86900850UG
Add to Cart
Add to Cart
Native, human growth hormone from pituitary gland. Single chain polypeptide hormone essential for growth of all tissues. Increases fat mobilization. Immunopotency: ≥5IU/mg (WHO 1st IRP).
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method 10mM sodium bicarbonate
Purification Method By weight
Quantity 50 μg
Immunogen Growth Hormone
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥97%, Iodination Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.