Description
- Polychromatic stain
- Used as stand alone stain or in combination with Wright Stain for staining of blood parasites such as malaria and rickettsia
Specifications
Specifications
| Quantity | 1L |
| Format | Liquid |
| Product Type | Giemsa Stain, Azure |