Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HARLECO™ Gram Stain Reagents

Catalog No. 23750027 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
500 mL
Product Type:
Decolorizer
Gram Iodine
Gram Stain Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
23-750027 500 mL Gram Iodine
23-750024 1 Set Gram Stain Kit
23-750028 500 mL Decolorizer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-750027 Supplier MilliporeSigma™ Supplier No. 65092D95

Complete set contains all reagents needed for Gram staining, or choose reagents individually.

 

  • Use qualitative technique for differential staining of bacteria from cultures and patient specimens by modified Hucker Gram stain method
  • Ideal for use with Midas III and other automated staining systems

 

Order Info

Individual reagents supplied in packs of four 500mL bottles

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mL
Format Bottle
For Use With (Application) Gram staining of bacteria
Stain Type Iodine
Product Type Gram Iodine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.