Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Harleco™ Phosphate Buffer

Catalog No. 23749912 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1L
4L
pH:
6.4
6.8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity pH
23-749912 1L 6.4
23-750090 4L 6.8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-749912 Supplier MilliporeSigma™ Supplier No. 121775

For use with the MIDAS™ III Automated Stainer

  • Used in Wright and Wright-Giemsa staining protocols

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1L
Boiling Point 100°C
Format Liquid
Color Colorless
pH 6.4
Product Type Phosphate Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.