Learn More
MilliporeSigma™ Hemocyanin, Keyhole Limpet, Megathura crenulata, BES/Mg2+ Buffer, Protein
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Concentration | Concentration on Label |
| Formulation | In 10mM MgSO4, 5mM BES buffer, 0.9% NaCl, pH 6.5. Sterile-filtered. |
| Purification Method | Size exclusion chromatography |
| Quantity | 250 mg |
| Source | Keyhole Limpet, Megathura crenulata |
| Immunogen | Hemocyanin |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Liquid |
| Purity or Quality Grade | ≥90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.