Learn More
MilliporeSigma™ Hemocyanin, Keyhole Limpet, Megathura crenulata, Glycerol, Protein
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Concentration | Concentration on Label |
| Formulation | In 10mM MgSO4, 5mM BES, 50% glycerol, 0.9% sodium chloride, pH 6.5. |
| Preparation Method | Further dilute with PBS or physiological saline. Material can be dialyzed to remove the glycerol or used as is for conjugation with glutaraldehyde, carbodiimide, or mixed anhydrides. |
| Purification Method | Size exclusion chromatography |
| Quantity | 1 g |
| Source | Keyhole Limpet, Megathura crenulata |
| Immunogen | Hemocyanin |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Liquid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.