Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hemocyanin, Keyhole Limpet, Megathura crenulata, Glycerol, Protein

Catalog No. 37481750MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-481-750MG 50 mg
37-481-7250MG 250 mg
37-481-71GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 37-481-750MG Supplier MilliporeSigma™ Supplier No. 37481750MG
Add to Cart
Add to Cart

Specifications

Concentration Concentration on Label
Formulation In 10mM MgSO4, 5mM BES, 50% glycerol, 0.9% sodium chloride, pH 6.5.
Preparation Method Further dilute with PBS or physiological saline. Material can be dialyzed to remove the glycerol or used as is for conjugation with glutaraldehyde, carbodiimide, or mixed anhydrides.
Purification Method Size exclusion chromatography
Quantity 50 mg
Source Keyhole Limpet, Megathura crenulata
Immunogen Hemocyanin
Regulatory Status RUO
Recombinant Native
Form Liquid
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.