Learn More
MilliporeSigma™ Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin-Free, Sterile-Liquid, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Concentration | Concentration on Label |
| Formulation | In 83mM sodium phosphate, 150mM NaCl, pH 7.2. |
| Purification Method | Size exclusion chromatography |
| Quantity | 25mg |
| Source | Keyhole Limpet, Megathura crenulata |
| Immunogen | Hemocyanin |
| Regulatory Status | RUO |
| Endotoxin Concentration | Endotoxin-Free |
| Recombinant | Native |
| Form | Liquid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.