Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hemocyanin, Keyhole Limpet, Megathura crenulata, Protein

Catalog No. 374805250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-480-5250MG 250 mg
37-480-51GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-480-5250MG Supplier MilliporeSigma™ Supplier No. 374805250MG
Add to Cart
Add to Cart
Stable solid containing more than 60% protein. Protein is 90% undissociated hemocyanin.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from BES buffer, MgSO4.
Purification Method Size exclusion chromatography
Quantity 250 mg
Source Keyhole Limpet, Megathura crenulata
Immunogen Hemocyanin
Storage Requirements Dissolve at 5 to 10mg/mL (protein) in PBS or other physiological buffer by GENTLE addition of the buffer (e.g. slowly added down the side of the tube). DO NOT vortex or agitate vigorously to dissolve product. (KLH is an extremely large protein complex and has a tendency to aggregate and precipitate from solutions.) For further dilutions, use an appropriate buffer and maintain the magnesium concentration at ≥10mM. Solutions should be stored in the refrigerator (4°C).
Regulatory Status RUO
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.