Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Heparin Cofactor II, Human Plasma, Protein

Catalog No. 375115100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-511-5100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-511-5100UG Supplier MilliporeSigma™ Supplier No. 375115100UG
Add to Cart
Add to Cart
Native heparin cofactor II from human plasma. Inhibits thrombin by forming a bimolecular complex in the presence of heparin.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 50% aqueous glycerol.
Purification Method SDS-PAGE
Quantity 100 μg
Immunogen Heparin Cofactor II
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.