Learn More
MilliporeSigma™ HPLC Solvent Filter/Degasser Filters
Catalog No. M6920011 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size (Metric):
0.2 μm
Catalog No. Pore Size (Metric)
M6920011 0.2 μm
1 options
Catalog No. M6920011 Supplier MilliporeSigma™ Supplier No. 6920011
PTFE replacement filter
EM Science™ HPLC Solvent Filter/Degasser Filters
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) To filter particulates that could damage expensive pumps and detectors, to remove dissolved oxygen in the solvent that could cause detector drift and instability
Type Replacement Filter
Pore Size (Metric) 0.2 μm
Membrane Material PTFE
Quantity 10 Pack