Learn More
Specifications
Specifications
| Name | Human IL-6 Recombinant Protein |
| Quantity | 20 μg |
| Source | E. coli |
| Recombinant | Recombinant |
| Purity or Quality Grade | Endotoxin level is less than 0.1ng/μg of protein |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.