Learn More
Specifications
Specifications
| Name | Human PDGF-BB Recombinant Protein, Animal Free |
| Quantity | 10 μg |
| Source | E. coli |
| Cross Reactivity | Cow, Monkey, Mouse, Pig, Rat |
| Recombinant | Recombinant |
| Purity or Quality Grade | Endotoxin free and greater than 98% pure by SDS-PAGE gel and HPLC analyses |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.