Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Human Recombinant ASF1a (H3/H4 chaperone/chromatin regulator)

Catalog No. 141114 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-111-4 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-111-4 Supplier MilliporeSigma™ Supplier No. 141114

Specifications

Characterization >95% purity by SDS PAGE
Concentration 0.5μg/μL
Formulation In buffer containing 20mM Tris-Cl pH 7.65, 0.15M NaCl, 10% glycerol and 1mM DTT
Name ASF1a (H3/H4 chaperone/chromatin regulator)
Purification Method Fast Protein Liquid Chromatography
Quantity 25 μg
Source E. coli
Storage Requirements -80°C for 6 months
Conjugate N-terminal GST-Tag
Cross Reactivity Human
Recombinant Recombinant
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.