Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Human Recombinant Histone H2A.Z

Catalog No. 141109 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-110-9 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-110-9 Supplier MilliporeSigma™ Supplier No. 141109
Add to Cart
Add to Cart

Human Histone H2A.Z Recombinant Protein

Human recombinant histone H2A.Z purified by Fast Protein Liquid Chromatography (FPLC) and verified by mass spectrometry

TRUSTED_SUSTAINABILITY

Specifications

Characterization >98% purity by SDS PAGE
Concentration 1 μg/μL
For Use With (Application) Chromatin Assembly Studies
Formulation Recombinant protein in a buffer containing 20 mM Tris-Cl pH 7.6, 1 mM EDTA, 0.15 M NaCl, 10% glycerol, 0.5 mM PMSF and 1 mM DTT
Name Histone H2A.Z
Purification Method Fast Protein Liquid Chromatography
Quantity 100 μg
Source E. coli
Storage Requirements -80°C for 12 months
Recombinant Recombinant
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.