Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Human TGF-α Recombinant Protein, Animal Free

Catalog No. GF313 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF313 Supplier MilliporeSigma™ Supplier No. GF313
Add to Cart
Add to Cart

Specifications

Name Human TGF-α Recombinant Protein, Animal Free
Quantity 100 μg
Source E. coli
Cross Reactivity Chicken, Mouse, Rat
Recombinant Recombinant
Purity or Quality Grade Endotoxin free and greater than 98% pure by SDS-PAGE gel and HPLC analyses
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.