Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hyaluronidase, Bovine Testes, Protein

Catalog No. 38593125KU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-593-125KU 25 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-593-125KU Supplier MilliporeSigma™ Supplier No. 38593125KU
Add to Cart
Add to Cart
Native hyaluronidase from bovine testes. Hyaluronic acid degrading endoglycosidase that catalyzes the hydrolysis of 1,4 linkages between N-acetylhexosamine and D-glucuronate residues in chondroitin, chondroitin-4-sulfate, chondroitin-6-sulfate, and hyaluronic acid.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Reconstitute in 20mM sodium phosphate, pH 6.9, 0.45% NaCl, and 0.01% BSA.
Quantity 25 kU
Immunogen Hyaluronidase
Storage Requirements Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C.
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.