Learn More
MilliporeSigma™ InnoCyte™ Cell Migration Assay, 24-well
Catalog No. CBA0171KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1/Pk.
Catalog No. Quantity
CBA0171KIT 1/Pk.
1 options
Catalog No. CBA0171KIT Supplier MilliporeSigma™ Supplier No. CBA0171KIT
Add to Cart
Add to Cart
Fluorometric Cell Migration Assay
TRUSTED_SUSTAINABILITY
Specifications
Description Cell Migration Assay
Detection Method Fluorometric
Includes 12 Sterile Cell Culture Inserts, Sterile 24-well Plate, Black Plate Strips, Cell Detachment Buffer, Fluorescent Dye, Latrunculin A (inhibitor), and a user protocol
Quantity 1/Pk.
Sample Type Adherent Cells