Specifications
| Description | Cell Migration Assay |
| Detection Method | Fluorometric |
| Includes | Sterile 96-Well Migration Chamber, Culture Tray, Cell Detachment Buffer, Fluorescent Dye, Latrunculin A (inhibitor), and a user protocol |
| Quantity | 1/Pk. |
| Sample Type | Adherent Cells |