Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ InnoCyte™ Monocyte Cell Migration Assay

Catalog No. CBA0981KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBA0981KIT 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBA0981KIT Supplier MilliporeSigma™ Supplier No. CBA0981KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains 96-Well monocyte cell migration chamber, black conical 96-well plate, D-PBS, Calcein-AM, and user protocol

  • Fluorometric cell migration assay.
  • Designed for monocyte motility studies; not for adherent cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Description Monocyte Cell Migration Assay
Detection Method Fluorometric
Includes 96-Well Monocyte Cell Migration Chamber, Black, Conical 96-Well Plate, D-PBS, Calcein-AM, and a user protocol
Quantity 1/Pk.
Sample Type Monocytes
Type Assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.