Learn More
MilliporeSigma™ Insulin, Zinc, Human, Recombinant Protein, P. pastoris
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Preparation Method | 0.01N HCl. Insulin is practically insoluble from pH 4.5-7.0. |
| Quantity | 50 mg |
| Source | P. pastoris |
| Immunogen | Insulin |
| Storage Requirements | Following reconstitution store in the refrigerator (4°C). Stock solutions are stable for up to 6 weeks at 4°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
| Form | Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.