Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ γ-Interferon, Rat, Lyophilized, Recombinant Protein, E. coli

Catalog No. 407321100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-732-1100UG 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-732-1100UG Supplier MilliporeSigma™ Supplier No. 407321100UG
Add to Cart
Add to Cart
Recombinant rat interferon-γ (IFN-γ) expressed in E. coli as a single, non-glycosylated polypeptide containing 135 amino acids. Purified by chromatographic methods. IFN-γ is a type II cytokine produced by activated T cells and natural killer cells. It plays a significant role immunoregulation and has been implicated in the induction of the expression of class II histocompatibility molecules in cells that do not normally express them, resulting in autoimmune disease.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.5X PBS, pH 7.2.
Preparation Method Reconstitute with sterile water to ≥100μg/mL, containing 0.1% HSA or BSA. Further dilute with aqueous buffers just prior to use.
Purification Method RP-HPLC
Quantity 100μg
Source E. coli
Immunogen γ-Interferon
Storage Requirements Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 7 days at 4°C or for up to 4 months at −20°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Species Rat
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.