Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Interleukin-2, Human, Recombinant Protein, E. coli

Catalog No. 40762310UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-762-310UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-762-310UG Supplier MilliporeSigma™ Supplier No. 40762310UG
Recombinant, human interleukin-2 expressed in E. coli. Has a variety of immunological functions. IL-2 is secreted by activated T cells. Acts as an autocrine agent to promote proliferation and maturation of activated T cells. Human IL-2 is active in murine cell lines. Exhibits about 60% amino acid homology with murine IL-2. Inhibits dexamethasone-induced apoptosis in T lymphocytes. Induces nitric oxide synthase.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Carrier-free.
Preparation Method Reconstitute to a concentration of 0.1-1mg/mL with sterile 100mM acetic acid. Further dilute in buffer or medium containing ≥0.1% BSA.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen Interleukin-2
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 3 months at −70°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.