Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Kallikrein, Human Plasma, Protein

Catalog No. 42030750UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-030-750UG 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-030-750UG Supplier MilliporeSigma™ Supplier No. 42030750UG
Add to Cart
Add to Cart
Native Kallikrein from human plasma. A serine protease that releases bradykinin from kininogen. Present in the plasma as an inactive precursor, prokallikrein.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In NaCl and Tris-HCl buffer.
Purification Method SDS-PAGE
Quantity 50μg
Immunogen Kallikrein
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.