Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Kininogen, High Molecular Weight, Single Chain, Human Plasma, Protein, Calbiochem

Catalog No. 4226861MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-268-61MG 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-268-61MG Supplier MilliporeSigma™ Supplier No. 4226861MG
Add to Cart
Add to Cart
Native, single chain, high molecular weight Kininogen from human plasma. Nonenzymatic cofactor of the contact activation system. Links prekallikrein to negatively charged surfaces, thereby allowing activation to kallikrein by surface-bound Factor α-XIIa. Also circulates in plasma in complex with Factor XI and may play a role in the activation of Factor XI to Factor XIa by Factor α-XIIa. Serves as a source of bradykinin, a potent vasoactive peptide important in hypotension studies.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 150mM NaCl, 4mM sodium acetate buffer, pH 5.3.
Purification Method SDS-PAGE
Quantity 1mg
Immunogen Kininogen
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Structural Form Single Chain, High Molecular Weight
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.