Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lactate Dehydrogenase, Rabbit Muscle, Protein

Catalog No. 42721725KU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-721-725KU 25 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-721-725KU Supplier MilliporeSigma™ Supplier No. 42721725KU
Add to Cart
Add to Cart
Native lactate dehydrogenase from rabbit muscle. Catalyzes the reversible conversion of pyruvate to lactate. The overall equilibrium of this reaction favors lactate formation.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method H2O or Low Ionic Strength Buffer
Quantity 25 kU
Immunogen Lactate Dehydrogenase
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Regulatory Status RUO
Species Rabbit
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.