Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Leptin, Human, Recombinant Protein, E. coli

Catalog No. 4297001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-970-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-970-01MG Supplier MilliporeSigma™ Supplier No. 4297001MG
Add to Cart
Add to Cart
Recombinant, human leptin expressed in E. coli. Native leptin is a product of the obese (ob) gene that serves as a ligand for the OB receptor (OB-R). Mice with mutations of the ob gene have been found to be obese and diabetic and to have reduced activity, metabolism, and body temperature. Reported to reduce hepatic glucose production by blocking phosphoenolpyruvate synthesis. Note: Following complete dissolution in 15mM HCl, add 7.5mM sterile NaOH and bring the pH to approximately 5.2.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a sterile filtered solution in PBS.
Purification Method SDS-PAGE
Quantity 1 mg
Source E. coli
Immunogen Leptin
Storage Requirements To reconstitute lyophilized leptin, add 150mM sterile HCl (0.5mL/1mg vial or 2.5mL/5mg vial) to the vial. After the protein is completely dissolved, add 7.5mM sterile NaOH (0.3mL/1mg vial or 1.5mL/5mg vial) to bring the pH to ∽5.2. Lyophilized samples are stable for at least six months at −70°C. Upon reconstitution, this cytokine can be stored under sterile conditions at 4°C for one month or at −70°C for three months without detectable loss of activity. Avoid repeated freeze/thaw cycles of reconstituted solutions.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.