Learn More
MilliporeSigma™ LiChroCART™ Column/Purospher™ STAR NH2 Sorbent HPLC Guard Cartridge
Catalog No. M1502670001 Shop All MilliporeSigma Products
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
Catalog No. Diameter (Metric) Inner
M1502670001 4 mm
1 options
Catalog No. M1502670001 Supplier MilliporeSigma™ Supplier No. 1502670001
Add to Cart
Add to Cart
HPLC guard cartridge
Purospher™ STAR NH2 HPLC columns are amino-bonded high-purity silica columns primarily for carbohydrate analysis. The particular benefits of these columns are the very high separation efficiency and the absence of metallic impurities, giving consistent symmetrical peaks.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.
Length (Metric) 4 mm
Type HPLC Guard Cartridge
Diameter (Metric) Inner 4 mm