Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lipocortin-I, HisTag™, Human, Recombinant Protein, E. coli

Catalog No. 43762150UG Shop All MilliporeSigma Products
Change view
Click to view available options
50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
43-762-150UG 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-762-150UG Supplier MilliporeSigma™ Supplier No. 43762150UG
Full-length, recombinant, human, lipocortin-I fused at the N-terminus to His·Tag™ and S·Tag™ sequences and expressed in E. coli. This preparation is qualified for use as a substrate for protein tyrosine kinases in in vitro assays. Lipocortin-I is a Ca2+/phospholipid-binding protein that promotes membrane fusion and exocytosis. It is known to be phosphorylated at Tyr20 by EGFR, Abl, and Src, at Thr215 by PKA, and at Thr23 and Ser26 by PKC. It is also a substrate for a variety of other protein tyrosine kinases.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, 0.2% Protease Inhibitor Cocktail Set VII.
Purification Method SDS-PAGE
Quantity 50μg
Source E. coli
Immunogen Lipocortin-I
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.