Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lipoproteins, High Density, Human Plasma, Protein

Catalog No. 43764110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-764-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-764-110MG Supplier MilliporeSigma™ Supplier No. 43764110MG
Add to Cart
Add to Cart
Native high density lipoproteins from human plasma. Cholesterol-carrier lipoprotein that acts as scavenger of tissue cholesterol. Important in cholesterol efflux from tissues. Involved in return of cholesterol from the periphery to the liver for removal as bile acids. Composition: 55-45% lipid, 45-55% protein.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 150mM NaCl, 0.01% EDTA, pH 7.4.
Purification Method Electrophoresis
Quantity 10 mg
Immunogen Lipoproteins
Storage Requirements This product does not contain preservatives, so it is recommended that it be used under a laminar flow hood. Following initial use, this product is stable for 1 week at 2-8°C.
Regulatory Status RUO
Structural Form High Density
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95% of total lipoprotein content
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.