Learn More
MilliporeSigma™ Upstate™ LSD1 (KDM1A), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set
Shop All MilliporeSigma ProductsDescription
Specifically detects LSD1 (KDM1A) Clone: in Human, Mouse, Rat samples, and it is validated for ChIP, Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | LSD1 (KDM1A) |
| Applications | ChIP Assay, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Anti-LSD1 (Rabbit Polyclonal). One vial containing 50μg affinity purified rabbit polyclonal in 72μL buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl and 0.05% sodium azide, before the addition of glycerol to 30%. Final concentration of antibody 0.7mg/mL. Normal rabbit IgG. One vial containing 125μg Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, PSA. One vial containing 75μL of 5μM of each primer specific for human prostate-specific antigen (PSA) enhancer region (chr19:51354179-51354263, hg19 build). FOR: GCC TGG ATC TGA GAG AGA TAT CAT C; REV: ACA CCT TTT TTT TTC TGG ATT GTT G |
| Gene Accession No. | O60341 |
| Gene Symbols | KDM1A, AOF2, KDM1, LSD1 |
| Host Species | Rabbit |
| Immunogen | Histidine-tagged recombinant protein corresponding to full length LSD1. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.