Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LX-2 Human Hepatic Stellate Cell Line

Catalog No. SCC064 Shop All MilliporeSigma Products
Change view
Click to view available options
LX-2 Human Hepatic Stellate Cell Line
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SCC064 LX-2 Human Hepatic Stellate Cell Line
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC064 Supplier MilliporeSigma™ Supplier No. SCC064

has been extensively characterized and retain key features of hepatic stellate cytokine signaling, neuronal gene expression, retinoid metabolism, and fibrogenesis , making them a highly suitable model of human hepatic fibrosis.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Stem cell culture and cell culture
Quantity 1 x 106cells/vial
Source Human hepatic stellate primary cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.