Learn More
MilliporeSigma™ MQuant™ pH-Box pH Indicator Papers with Acilit, Neutralit, Alkalit, pH 0.5 to 13
Shop All MilliporeSigma Products

Description
Includes
Color scale
Suited for all media in environmental analysis and industrial in-process controls.
- Rapid on-the-spot determination
- Acilit covers range 0.5 to 5.0 pH
- Neutralit covers range 5.5 to 9.0 pH
- Alkalit covers 9.0 to 13.0 pH
- Biologically degradable polyester foil and low reagent content in reaction zones support problem-free waste disposal
- Range: 0.5 to 13.0
- Graduations: 0.5-1.0-1.5-2.0-2.5-3.0-3.5-4.0-4.5-5.0-5.5-6.0-6.5-7.0-7.5-8.0-8.5-9.0-9.5-10.0-10.5-11.0-11.5-12.0-12.5-13.0
Order Info
Three rolls of 4.8m each
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| pH Range | 0.5 to 13 |
| For Use With (Application) | Suited for all media in environmental analysis and industrial in-process controls |
| Includes | Color scale |
| Quantity | 3 Rolls |
| Type | pH Box pH Indicator Paper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.