Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ pH Indicator Congo Red Papers

Catalog No. M1095140003 Shop All MilliporeSigma Products
Change view
Click to view available options
pH Range:
<3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
M1095140003 <3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1095140003 Supplier MilliporeSigma™ Supplier No. 1.09514.0003
Add to Cart
Add to Cart

Measure pH swiftly without using instruments

Suited for all media in environmental analysis and industrial in-process controls.

  • High-quality filter paper impregnated with iindicator solutions
  • Rolls better suited for long-term storage — protected against external influences
  • Biologically degradable polyester foil and low reagent content in reaction zones support problem-free waste disposal
  • Reagent Ph Eur Grade

Range: pH less than 3, blue-violet/greater than 5, red-orange

Order Info

Pack of three rolls (4.8m)

TRUSTED_SUSTAINABILITY

Specifications

Color Indicator Blue to Violet
pH Range <3
For Use With (Application) For long term storage, since they are protected against external influences
Quantity 3 Rolls
Type MColorpHast™ pH Indicator Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.