Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ pH Indicator Papers
GSA_VA

Catalog No. M1095570003 Shop All MilliporeSigma Products
Change view
Click to view available options
pH Range:
6.4 to 8
Graduations:
6.4 to >8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range Graduations
M1095570003 6.4 to 8 6.4 to >8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1095570003 Supplier MilliporeSigma™ Supplier No. 1.09557.0003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Measure pH onsite without instruments or extensive preparation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

pH Range 6.4 to 8
For Use With (Application) Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
Graduations 6.4 to >8
Quantity 3 x 4.8 Rolls
Storage Requirements 10°C to 25°C
Type Special pH Indicator Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.