Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ pH Indicator Strips, pH 2.0 to 9.0

Catalog No. M1095020001 Shop All MilliporeSigma Products
Change view
Click to view available options
pH Range:
2 to 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
M1095020001 2 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1095020001 Supplier MilliporeSigma™ Supplier No. 1.09502.0001
Add to Cart
Add to Cart

Includes

Color scale

Measure pH swiftly without using instruments

Special indicator for turbid solutions/suspensions

  • Range: pH 2.0 to 9.0
  • Graduations: 2.0-3.0-4.0-5.0-6.0--7.0-8.0-9.0

Order Info

Pack of 100 tests

TRUSTED_SUSTAINABILITY

Specifications

pH Range 2 to 9
For Use With (Application) For pH measurements in turbid solutions, environmental analysis/process control
Quantity Each
Type MColorpHast™ pH Indicator Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.