Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ pH Test Strips and Indicator Papers

Catalog No. M1095350001 Shop All MilliporeSigma Products
Change view
Click to view available options
pH Range:
0 to 14
0 to 2.5
0 to 6
0.5 to 5
11 to 13
2 to 9
2.5 to 4.5
4 to 7
5 to 10
6.5 to 10
7.5 to 14
9.5 to 13
Type:
Acilit pH Indicator Paper
Acilit pH Indicator Strip
Neutralit pH Indicator Strip
Special pH Indicator Strip
Standard pH Indicator Strip
Universal pH Indicator Strip
Quantity:
1 Box
1 Box of 100
100 Box
100 Strips
100 Strips in Plastic Box
100 Strips/Ea.
100 Test Strips
3 x 4.8 Rolls
6 Box
6 Pack
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range Type Quantity
M1095350001 0 to 14 Universal pH Indicator Strip 100 Strips
M1095330001 5 to 10 Neutralit pH Indicator Strip 100 Strips
M1095310001 0 to 6 Acilit pH Indicator Strip 100 Strips/Ea.
M1095320001 7.5 to 14 Acilit pH Indicator Strip 100 Strips in Plastic Box
M1095400001 0 to 2.5 Special pH Indicator Strip 100 Box
M1095410001 2.5 to 4.5 Special pH Indicator Strip 1 Box
M1095430001 6.5 to 10 Special pH Indicator Strip 1 Box of 100
M1095420001 4 to 7 Special pH Indicator Strip 100 Box
M1095620003 9.5 to 13 Acilit pH Indicator Paper 3 x 4.8 Rolls
M1095450001 11 to 13 Special pH Indicator Strip 1 Box
M1095840001 2 to 9 Standard pH Indicator Strip 100 Test Strips
M1095330007 5 to 10 Neutralit pH Indicator Strip 6 Pack
M1095310007 0 to 6 Acilit pH Indicator Strip 6 Pack
M1095350007 0 to 14 Universal pH Indicator Strip 6 Pack
M1095420007 4 to 7 Special pH Indicator Strip 6 Box
M1095430007 6.5 to 10 Special pH Indicator Strip 6 Box
M1095600003 0.5 to 5 Acilit pH Indicator Paper 3 x 4.8 Rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. M1095350001 Supplier MilliporeSigma™ Supplier No. 1.09535.0001
Add to Cart
Add to Cart

Measure pH onsite without instruments or extensive preparation

Fit into a pocket and provide easy, rapid results—suited to all media in environmental analysis and industrial in-process controls.

  • Differentiated color gradations for a more precise evaluation
  • Traditional indicator paper or MQuant™ nonbleeding universal strips available
Traditional strips
  • High-quality papers impregnated with indicator or mixed-indicator solutions, cut to size and dried
  • Supplied in a roll format that protects strips against moisture, light or ambient gases for longer storage life
Plastic indicator strips provide differentiated color gradations for a more precise evaluation
  • Special indicator dyes covalently bonded to reagent papers
  • Indicator does not bleed, so strips will not contaminate the medium, even if left for long periods of time
Long shelf life
  • Can be used for three to five years when stored at 10 to 25°C in a light-protected area
  • Reseal quickly after each use
Consistently high quality
  • Tests checked and calibrated using certified buffer solutions
  • Buffer solutions are traceable to primary reference materials from NIST™ and PTB

Order Info

Packaging: indicator papers come in lengths of 4.8m in roll dispenser; MQuant™ special indicator pH strips come as strips packed in boxes of 100 tests each.

TRUSTED_SUSTAINABILITY

Specifications

pH Range 0 to 14
For Use With (Application) Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
Graduations 15
Quantity 100 Strips
Storage Requirements 10°C to 25°C
Type Universal pH Indicator Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.