Learn More
Description
Fit into a pocket and provide easy, rapid results—suited to all media in environmental analysis and industrial in-process controls.
- Differentiated color gradations for a more precise evaluation
- Traditional indicator paper or MQuant™ nonbleeding universal strips available
- High-quality papers impregnated with indicator or mixed-indicator solutions, cut to size and dried
- Supplied in a roll format that protects strips against moisture, light or ambient gases for longer storage life
- Special indicator dyes covalently bonded to reagent papers
- Indicator does not bleed, so strips will not contaminate the medium, even if left for long periods of time
- Can be used for three to five years when stored at 10 to 25°C in a light-protected area
- Reseal quickly after each use
- Tests checked and calibrated using certified buffer solutions
- Buffer solutions are traceable to primary reference materials from NIST™ and PTB
Order Info
Packaging: indicator papers come in lengths of 4.8m in roll dispenser; MQuant™ special indicator pH strips come as strips packed in boxes of 100 tests each.
Specifications
Specifications
| pH Range | 4 to 7 |
| For Use With (Application) | Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination |
| Graduations | 10 |
| Quantity | 100 Box |
| Storage Requirements | 10°C to 25°C |
| Type | Special pH Indicator Strip |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.