Learn More
MilliporeSigma™ MQuant™ Chlorine Test Kits
Catalog No. M1149780001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Chlorine Test Kit
Chlorine Test Kit (Liquid)
Chlorine Test Kit Refill
Cl Test Kit (Liquid) Refill
Range:
0.01 to 0.3 mg/L
0.1 to 2 mg/L Cl2
0.25 to 15 mg/L Cl2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range
M1149780001 Chlorine Test Kit (Liquid) 0.1 to 2 mg/L Cl2
M1149770002 Chlorine Test Kit Refill 0.25 to 15 mg/L Cl2
M1149790002 Cl Test Kit (Liquid) Refill 0.1 to 2 mg/L Cl2
M1144340001 Chlorine Test Kit 0.01 to 0.3 mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1149780001 Supplier MilliporeSigma™ Supplier No. 1149780001
Add to Cart
Add to Cart
Can be used for the determination of chlorine.
The liquid reagent can be used for the determination of free and total chlorine with the MQuant Chlorine test. The chlorine concentration can be measured semiquantitatively by visual comparison of the colour of the measurement solution with the colour fields of the colour disk.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 600 Tests
Range 0.1 to 2 mg/L Cl2
Type Chlorine Test Kit (Liquid)