Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ MED1, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710530MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-105-30MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-105-30MI Supplier MilliporeSigma™ Supplier No. 1710530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects MED1 Clone: in Human, Mouse samples, and it is validated for ChIP, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MED1
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-MED1 (Rabbit Polyclonal). One vial containing 50μg affinity purified rabbit polyclonal in 72μL buffer containing Tris-citrate/phosphate buffer (pH 7 to 8) and 0.09% sodium azide, before the addition of glycerol to 30% (0.7mg/mL final). Normal Rabbit IgG. One vial containing 125μg of purified Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, PSA. One vial containing 75μL of 5μM of each primer specific for human prostate-specific antigen enhancer region (chr19:51354179-51354263, hg19 build). FOR: GCC TGG ATC TGA GAG AGA TAT CAT C; REV: ACA CCT TTT TTT TTC TGG ATT GTT G
Gene Accession No. Q15648
Gene Symbols ARC2052, CRSP1, CRSP200, DRIP205, DRIP230, PBP, PPARBP, PPARGBP, RB18A, TRAP220, TRIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to human MED1 (a.a. 1525-1581).
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_004765
Includes Use ChIPAb+ MED1 ChIP Validated mouse monoclonal antibody for the detection of MED also known as Mediator of RNA polymerase II transcription subunit 1 and CRSP1/TRAP220, includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.