Learn More
Description
Specifications
Specifications
| Purification Method | HPLC |
| Purity | ≥95% |
| Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 months at −20°C. |
| Format | Lyophilized |
| Formulation | Supplied as an acetate salt. Sold on the basis of peptide content. |
| Product Type | α-Melanocyte-Stimulating Hormone |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.