Learn More
MilliporeSigma™ Microcrystalline Cellulose Loose Sorbent for TLC
Catalog No. M1023300500 Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging Quantity:
500 g
Catalog No. Packaging Quantity
M1023300500 500 g
1 options
Catalog No. M1023300500 Supplier MilliporeSigma™ Supplier No. 1.02330.0500
Add to Cart
Add to Cart
for thin-layer chromatography.
CAS 9004-34-6, pH 5-7.5 (100g/l, H2O, 20°C)(slurry). Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40μm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
TRUSTED_SUSTAINABILITY
Specifications
Type Cellulose Microcrystalline
Packaging Quantity 500 g