Learn More
MilliporeSigma™ Microfil™ S Filtration Devices
Catalog No. MVHAWGS24 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.45 μm
Catalog No. Pore Size
MVHAWGS24 0.45 μm
1 options
This item is not returnable. View return policy
Catalog No. MVHAWGS24 Supplier MilliporeSigma™ Supplier No. MVHAWGS24
This item is not returnable. View return policy
Fast, dependable alternative to conventional multi-step methods
Devices consist of a funnel, protective lid, and filtration support so routine microbiological testing is fast and easy.
  • Ensure consistent recovery of microorganisms
  • Smooth, hydrophobic funnel surface repels aqueous sample residues to ensure that microorganisms are collected on the membrane, not on funnel walls
  • Convex support shape eliminates pleating and distortion of membrane due to expansion
  • Three membrane selections offer versatility for your specific application
Benefits
  • Presterilized, individually packaged No membrane handling prior to filtering
  • No filter holders, funnels and attachments to assemble and sterilize
  • No sterilizing the filter support after each filtration Protective lid reduces external contamination risk Pre-assembled to speed sampling and shorten preparation Easy-to-remove funnel and membrane reduce test time
Compliance
Comply with USP/EP Guidelines. Certificate of Quality demonstrates that the device has been quality-control tested with Pharmacopoeia organisms and demonstrates acceptable growth.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) EMD Millipore Microfil S Filtration Devices
Material (Membrane) MCE
Pore Size 0.45 μm