Learn More
MilliporeSigma™ Millicup™ Self-Contained, Disposable, Vacuum Bottle-Top Filter for HPLC
Shop All MilliporeSigma ProductsDescription
Features & Benefits:
- No preparation or clean-up saves time
- Available with low-protein binding hydrophilic Durapore PVDF, hydrophobic Fluoropore PTFE, and hydrophilic LCR PTFE membranes
- Ultraclean design and safe operation
Applications: Mobile Phase Preparation, Buffer Filtration
Specifications
Specifications
| Diameter (Metric) Inner | 8.4 cm |
| Height (English) | 4.7 in. |
| Height (Metric) | 12 cm |
| Pore Size | 0.45 μm |
| Membrane Material | Hydrophobic PTFE |
| For Use With (Application) | HPLC |
| Type | Bottle Top Filter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.