Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millicup™ Self-Contained, Disposable, Vacuum Bottle-Top Filter for HPLC
GSA_VA

Catalog No. SJLHM4710 Shop All MilliporeSigma Products
Change view
Click to view available options
Membrane Material:
Durapore PVDF
Hydrophilic PTFE
Hydrophobic PTFE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Membrane Material
SJLHM4710 Hydrophilic PTFE
SJHVM4710 Durapore PVDF
SJFHM4710 Hydrophobic PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SJLHM4710 Supplier MilliporeSigma™ Supplier No. SJLHM4710
Add to Cart
Add to Cart

Non sterile Universal bottletop vacuum filter

The Millicup Filter Unit provides fast and effective filtration of solvents and buffers (up to 3L) used in HPLC and other analytical techniques.Features & Benefits:•No preparation or clean-up saves time •Available with low-protein binding hydrophilic Durapore PVDF, hydrophobic Fluoropore PTFE, and hydrophilic LCR PTFE membranes •Ultraclean design and safe operationApplications:Mobile Phase Preparation, Buffer Filtration

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 8.4 cm
Height (English) 4.7 in.
Height (Metric) 12 cm
Pore Size 0.45 μm
Membrane Material Hydrophilic PTFE
For Use With (Application) HPLC
Type Bottle Top Filter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.