Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millipak™ Disposable Filter Units

Catalog No. MPHL06GF2 Shop All MilliporeSigma Products
Change view
Click to view available options
Connectors:
NPTM (In), SHB with Bell (Out)
SHB (In), SHB with Bell (Out)
Sanitary Flange (In/Out)
Flow Rate:
0.7 L/min. at 1.75 bar
1.1 L/min. at 1.75 bar
1.5 L/min. at 1.75 bar
1.9 L/min. at 1.75 bar
10.5 L/min. at 2.45 bar
2.8 L/min. at 1.75 bar
3.8 L/min. at 1.75 bar
6.4 L/min. at 2.45 bar
7.6 L/min. at 2 bar
8.5 L/min. at 2.45 bar
Volume (Metric):
10 L
20 L
30 L
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connectors Flow Rate Volume (Metric)
MPHL06GF2 Sanitary Flange (In/Out) 10.5 L/min. at 2.45 bar 30 L
MPGL02GF2 Sanitary Flange (In/Out) 1.5 L/min. at 1.75 bar 10 L
MPGL02GH2 SHB (In), SHB with Bell (Out) 1.5 L/min. at 1.75 bar 20 L
MPGL04GH2 SHB (In), SHB with Bell (Out) 2.8 L/min. at 1.75 bar 20 L
MPGL04GF2 Sanitary Flange (In/Out) 2.8 L/min. at 1.75 bar 20 L
MPGL06CB1 NPTM (In), SHB with Bell (Out) 7.6 L/min. at 2 bar 30 L
MPGL06GF2 Sanitary Flange (In/Out) 7.6 L/min. at 2 bar 30 L
MPGL06GH2 SHB (In), SHB with Bell (Out) 3.8 L/min. at 1.75 bar 30 L
MPHL06GH2 SHB (In), SHB with Bell (Out) 10.5 L/min. at 2.45 bar 30 L
MPHL04GH2 SHB (In), SHB with Bell (Out) 8.5 L/min. at 2.45 bar 20 L
MPHL02GH2 SHB (In), SHB with Bell (Out) 6.4 L/min. at 2.45 bar 10 L
MPVL02GH2 SHB (In), SHB with Bell (Out) 0.7 L/min. at 1.75 bar 10 L
MPVL06GH2 SHB (In), SHB with Bell (Out) 1.9 L/min. at 1.75 bar 30 L
MPVL04GH2 SHB (In), SHB with Bell (Out) 1.1 L/min. at 1.75 bar 20 L
MPGL04GK2 NPTM (In), SHB with Bell (Out) 2.8 L/min. at 1.75 bar 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. MPHL06GF2 Supplier MilliporeSigma™ Supplier No. MPHL06GF2
Add to Cart
Add to Cart

Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.

Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.

  • Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
  • Point-of-use filter for high-purity water
  • Ideally suited for high value-added applications
  • Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
  • Presterilized units are gamma irradiated
  • Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
  • 100% integrity tested to guarantee integrity

TRUSTED_SUSTAINABILITY

Specifications

Connectors Sanitary Flange (In/Out)
Disposable Single-use
Material Polycarbonate
Material (Membrane) PVDF
Pore Size 0.45 μm
Sterility Sterile
Filtration Area 300 cm2
Flow Rate 10.5 L/min. at 2.45 bar
For Use With (Application) Removal of particles and microorganisms from liquids and gases
Inlet Connection (English) 0.75 in.
Inlet Connection (Metric) 19.05 mm
Inlet Style Sanitary Flange
Max. Inlet Pressure 75 psi at 25°C
No. per Pack 2/Pk.
Outlet Fitting 0.75 in. (19.05 mm)
Outlet Style Sanitary Flange
Type Filter Unit
Volume (Metric) 30 L
Length (English) 3.7 in.
Length (Metric) 9.4 cm
Diameter (English) 3 in.
Diameter (Metric) 7.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.